KEY465193773 Key Organics / BIONET

5-oxo-4-{[3-(trifluoromethyl)phenyl]methyl}-4H,5H-pyrazolo[1,5-a]quinazoline-3-carbonitrile

  • MDL number MFCD06496150
  • Cas Number : 861208-18-6
  • Molecular Weight : 368.319

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66523

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1Y-0826_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart