KEY465193774 Key Organics / BIONET

methyl 4-cyano-3-(2,5-dimethyl-1H-pyrrol-1-yl)thiophene-2-carboxylate

  • MDL number MFCD06496152
  • Cas Number : 900019-87-6
  • Molecular Weight : 260.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66527

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1Y-0830_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart