KEY465193778 Key Organics / BIONET

4,6-dimethyl-2-[4-(pyridine-2-carbonyl)-1H-1,2,3-triazol-1-yl]pyrimidine

  • MDL number MFCD08064718
  • Cas Number : 923106-19-8
  • Molecular Weight : 280.291

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66537

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1Y-0847_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart