KEY465193786 Key Organics / BIONET

2-amino-4-(4-methylphenyl)-4H-benzo[g]chromene-3-carbonitrile

  • MDL number MFCD06496163
  • Cas Number : 861208-30-2
  • Molecular Weight : 312.372

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66558

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1Y-0962_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart