KEY465193787 Key Organics / BIONET

(2E)-3-(2H-1,3-benzodioxol-5-yl)-2-(thiophen-2-yl)prop-2-enenitrile

  • MDL number MFCD00174346
  • Cas Number : 1002342-12-2
  • Molecular Weight : 255.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66565

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1Y-5051_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart