KEY465193794 Key Organics / BIONET

1,1-dimethyl-2-[(Z)-1-(methylsulfanyl)-2-nitroethenyl]hydrazine

  • MDL number MFCD00138258
  • Cas Number : 120739-61-9
  • Molecular Weight : 177.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66590

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2B-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart