KEY465193797 Key Organics / BIONET

(2E)-2-cyano-N-(2-fluoro-4-hydroxyphenyl)-3-(4-methoxyphenyl)prop-2-enamide

  • MDL number MFCD00127295
  • Cas Number : 338393-98-9
  • Molecular Weight : 312.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66596

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2B-009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart