KEY465193811 Key Organics / BIONET

5-[(benzylamino)methylidene]-1,3-dimethyl-1,3-diazinane-2,4,6-trione

  • MDL number MFCD00127313
  • Cas Number : 338394-14-2
  • Molecular Weight : 273.292

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66627

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2B-096_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart