KEY465193812 Key Organics / BIONET

2-oxo-2-(3-oxopyrazolidin-1-yl)ethyl pyrrolidine-1-carbodithioate

  • MDL number MFCD00127314
  • Cas Number : 338394-15-3
  • Molecular Weight : 273.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66628

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2B-099_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart