KEY465193815 Key Organics / BIONET

3-(2,6-dichlorophenyl)-5-[(E)-2-(phenylamino)ethenyl]-1,2-oxazole-4-carbonitrile

  • MDL number MFCD00127319
  • Cas Number : 338394-19-7
  • Molecular Weight : 356.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66633

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2C-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart