KEY465193816 Key Organics / BIONET

5-[(E)-2-[(4-chlorophenyl)amino]ethenyl]-3-(2,6-dichlorophenyl)-1,2-oxazole-4-carbonitrile

  • MDL number MFCD00127320
  • Cas Number : 338394-20-0
  • Molecular Weight : 390.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66634

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2C-004_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart