KEY465193825 Key Organics / BIONET

2,2-bis(ethylsulfanyl)-N-({[3-(trifluoromethyl)phenyl]carbamothioyl}amino)acetamide

  • MDL number MFCD00138464
  • Cas Number : 338394-35-7
  • Molecular Weight : 397.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66654

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2C-044_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart