KEY465193826 Key Organics / BIONET

N-methyl-5-nitro-2-(pyridin-4-yl)pyrimidin-4-amine

  • MDL number MFCD00138466
  • Cas Number : 338394-37-9
  • Molecular Weight : 231.215

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66656

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2C-053_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart