KEY465193831 Key Organics / BIONET

N'-[(1E)-1-amino-2-cyanoeth-1-en-1-yl]benzohydrazide

  • MDL number MFCD00172126
  • Cas Number : 136773-45-0
  • Molecular Weight : 202.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66684

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2D-022_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart