KEY465193835 Key Organics / BIONET

N-methyl-4-nitro-1-[4-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

  • MDL number MFCD00127334
  • Cas Number : 321533-61-3
  • Molecular Weight : 286.214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66703

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2D-052_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart