KEY465193871 Key Organics / BIONET

2-{3-[(2,4-dichlorophenoxy)methyl]-1H-1,2,4-triazol-5-yl}acetonitrile

  • MDL number MFCD00172025
  • Cas Number : 338395-57-6
  • Molecular Weight : 283.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66835

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2F-024_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart