KEY465193877 Key Organics / BIONET

N'-[(1Z)-1-amino-2-(2,4-dichlorophenoxy)ethylidene]-4-chlorobenzohydrazide

  • MDL number MFCD00142081
  • Cas Number : 338395-78-1
  • Molecular Weight : 372.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66861

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-052_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart