KEY465193880 Key Organics / BIONET

methyl 1-(4-methoxyphenyl)-6-oxo-4-(pyridin-2-ylsulfanyl)-1,6-dihydropyridazine-3-carboxylate

  • MDL number MFCD00141159
  • Cas Number : 338396-07-9
  • Molecular Weight : 369.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66879

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-073_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart