KEY465193882 Key Organics / BIONET

(2E)-4-[1-methyl-5-(2,2,2-trichloroacetyl)-1H-pyrrol-3-yl]-4-oxobut-2-enoic acid

  • MDL number MFCD00141859
  • Cas Number : 338395-79-2
  • Molecular Weight : 324.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66881

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-075_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart