KEY465193886 Key Organics / BIONET

methyl 7-amino-2-(4-methoxyphenyl)-3-oxo-2H,3H-thieno[3,2-c]pyridazine-6-carboxylate

  • MDL number MFCD00141165
  • Cas Number : 338395-94-1
  • Molecular Weight : 331.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66895

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-092_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart