KEY465193888 Key Organics / BIONET

3-amino-5-(4-methoxyphenyl)-1H,5H,6H-pyrazolo[4,3-c]pyridazin-6-one

  • MDL number MFCD00141168
  • Cas Number : 338395-98-5
  • Molecular Weight : 257.253

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66898

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-098_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart