KEY465193889 Key Organics / BIONET

methyl 6-oxo-4-phenoxy-1-[3-(trifluoromethyl)phenyl]-1,6-dihydropyridazine-3-carboxylate

  • MDL number MFCD00141169
  • Cas Number : 338396-00-2
  • Molecular Weight : 390.318

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66899

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-099_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart