KEY465193903 Key Organics / BIONET

1,5-diethyl (2E,4E)-4-[amino(4-phenylpiperazin-1-yl)methylidene]-2-cyanopent-2-enedioate

  • MDL number MFCD00140598
  • Cas Number : 338396-62-6
  • Molecular Weight : 398.463

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67027

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-930_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart