KEY465193904 Key Organics / BIONET

3-imino-3-[4-(4-methoxyphenyl)piperazin-1-yl]propanenitrile

  • MDL number MFCD00664736
  • Cas Number : 338396-63-7
  • Molecular Weight : 258.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67028

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-932_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart