KEY465193905 Key Organics / BIONET

ethyl (2Z,4E)-5-amino-2,4-dicyano-5-[4-(4-methoxyphenyl)piperazin-1-yl]penta-2,4-dienoate

  • MDL number MFCD00140599
  • Cas Number : 338396-64-8
  • Molecular Weight : 381.436

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67030

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-935_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart