KEY465193906 Key Organics / BIONET

5-(4-chlorophenyl)-2-[4-(4-methoxyphenyl)piperazin-1-yl]-1H-pyrrole-3-carbonitrile

  • MDL number MFCD00171775
  • Cas Number : 338396-65-9
  • Molecular Weight : 392.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67031

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-936_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart