KEY465193909 Key Organics / BIONET

8-benzyl-2-(hydroxymethyl)-octahydro-[1,2]oxazolo[2,3-a]pyridin-8-ium bromide

  • MDL number MFCD01936156
  • Cas Number : 119204-20-5
  • Molecular Weight : 328.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67047

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-959_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart