KEY465193911 Key Organics / BIONET

3,5-diethyl 2-oxo-6-(pyrrolidin-1-yl)-1,2-dihydropyridine-3,5-dicarboxylate

  • MDL number MFCD00202515
  • Cas Number : 128099-66-1
  • Molecular Weight : 308.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67052

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2F-980_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart