KEY465193913 Key Organics / BIONET

1-{3-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1,2-oxazol-5-yl}ethan-1-one

  • MDL number MFCD01315015
  • Cas Number : 477872-99-4
  • Molecular Weight : 290.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67061

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2G-014_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart