KEY465193921 Key Organics / BIONET

3-nitro-4-(1H-pyrazol-1-yl)-N-[3-(trifluoromethyl)phenyl]benzamide

  • MDL number MFCD00141988
  • Cas Number : 321534-59-2
  • Molecular Weight : 376.295

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67117

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2G-342S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart