KEY465193923 Key Organics / BIONET

N-(4-methoxyphenyl)-3-nitro-4-(1H-pyrazol-1-yl)benzamide

  • MDL number MFCD00141990
  • Cas Number : 321534-61-6
  • Molecular Weight : 338.323

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67122

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2G-348S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart