KEY465193929 Key Organics / BIONET

N,N-dimethyl-6-nitro-1,2-benzoxazole-3-carboxamide

  • MDL number MFCD00665466
  • Cas Number : 400079-09-6
  • Molecular Weight : 235.199

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67153

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2G-405S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart