KEY465193931 Key Organics / BIONET

methyl 4-[4-(3,4-dichlorophenyl)-1,3-thiazol-2-yl]benzoate

  • MDL number MFCD00171794
  • Cas Number : 338397-23-2
  • Molecular Weight : 364.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67169

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2G-430S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart