KEY465193943 Key Organics / BIONET

N-[(E)-2-(benzenesulfonyl)-2-nitroethenyl]-4-(trifluoromethyl)aniline

  • MDL number MFCD00214856
  • Cas Number : 338397-54-9
  • Molecular Weight : 372.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67217

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2H-018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart