KEY465193947 Key Organics / BIONET

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-N-(prop-2-yn-1-yl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD00231934
  • Cas Number : 338397-74-3
  • Molecular Weight : 327.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67237

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2H-043_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart