KEY465193954 Key Organics / BIONET

ethyl (2E)-2-[2-(4-chloro-2-nitrophenyl)hydrazin-1-ylidene]-2-acetamidoacetate

  • MDL number MFCD04124862
  • Cas Number : 861208-39-1
  • Molecular Weight : 328.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67262

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2H-302S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart