KEY465193964 Key Organics / BIONET

5-methyl-2-(pyridin-4-yl)-1,3-thiazol-4-yl benzoate

  • MDL number MFCD00172525
  • Cas Number : 338398-94-0
  • Molecular Weight : 296.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67384

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2H-458S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart