KEY465193969 Key Organics / BIONET

5-methyl-2-(thiophen-2-yl)-1,3-thiazol-4-ol

  • MDL number MFCD00172531
  • Cas Number : 131786-51-1
  • Molecular Weight : 197.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67397

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2H-473S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart