KEY465193971 Key Organics / BIONET

5-methyl-2-(pyridin-3-yl)-1,3-thiazol-4-yl 4-chlorobenzoate

  • MDL number MFCD00172533
  • Cas Number : 338399-00-1
  • Molecular Weight : 330.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67399

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2H-476S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart