KEY465193982 Key Organics / BIONET

5-acetyl-1-(2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD00172969
  • Cas Number : 338399-27-2
  • Molecular Weight : 376.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67434

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2H-919_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart