KEY465193983 Key Organics / BIONET

5-acetyl-3-[(4-fluorophenyl)methyl]-1-(4-methylpiperazin-1-yl)-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD00172970
  • Cas Number : 338399-28-3
  • Molecular Weight : 360.389

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67435

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2H-920_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart