KEY465193990 Key Organics / BIONET

1-(2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)-3-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

  • MDL number MFCD00172990
  • Cas Number : 338770-14-2
  • Molecular Weight : 373.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67492

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2H-980_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart