KEY465193998 Key Organics / BIONET

1,1-bis(4-fluorophenyl)-2-{[(2-methoxyphenyl)methyl]amino}ethan-1-ol

  • MDL number MFCD00231466
  • Cas Number : 338771-02-1
  • Molecular Weight : 369.412

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67547

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2J-307S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart