KEY465194002 Key Organics / BIONET

1,1-bis(4-fluorophenyl)-2-(4-methylpiperazin-1-yl)ethan-1-ol hydrochloride

  • MDL number MFCD00231473
  • Cas Number : 317822-08-5
  • Molecular Weight : 368.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67570

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2J-332S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart