KEY465194005 Key Organics / BIONET

2-{[(2,4-dichlorophenyl)methyl]amino}-1-(4-fluorophenyl)-1-phenylethan-1-ol

  • MDL number MFCD00231481
  • Cas Number : 338771-51-0
  • Molecular Weight : 390.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67581

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2J-345S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart