KEY465194008 Key Organics / BIONET

ethyl 2-{[5-methoxy-2-(pyridin-2-yl)pyrimidin-4-yl]sulfanyl}acetate

  • MDL number MFCD00215582
  • Cas Number : 338771-59-8
  • Molecular Weight : 305.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67588

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2J-353S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart