KEY465194010 Key Organics / BIONET

ethyl 2-{[5-methoxy-2-(pyridin-2-yl)pyrimidin-4-yl]sulfanyl}propanoate

  • MDL number MFCD00215586
  • Cas Number : 338771-77-0
  • Molecular Weight : 319.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67602

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2J-373S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart