KEY465194015 Key Organics / BIONET

N2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-N1-(2-nitrophenyl)ethane-1,2-diamine

  • MDL number MFCD00793063
  • Cas Number : 338772-75-1
  • Molecular Weight : 360.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67713

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2J-935_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart