KEY465194017 Key Organics / BIONET

4-{2-[(4-nitrophenyl)sulfanyl]-4,5-dihydro-1H-imidazol-1-yl}benzoic acid

  • MDL number MFCD00664199
  • Cas Number : 338772-90-0
  • Molecular Weight : 343.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67723

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2J-947_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart