KEY465194018 Key Organics / BIONET

2-phenyl-9,12-dioxa-1,2,4-triazadispiro[4.2.4^{8}.2^{5}]tetradecan-3-one

  • MDL number MFCD00974195
  • Cas Number : 338773-04-9
  • Molecular Weight : 289.335

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67730

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2J-956_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart